Chloroform ≥99.8% stabilised, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™

Supplier: HONEYWELL
Danger


Stabilization: Stabilised with ethanol ~1%

25669-1L 25669-2.5L 25669-6X1L
25669-1LEA 88.3 EUR
25669-1L 25669-2.5L 25669-6X1L
Chloroform ≥99.8% stabilised, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Chloroform

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent


Beilstein Number: 1731042

Formula: CHCl₃
Boiling Pt: 62 °C
Melting Pt: –64 °C
Density: 1,49 g/cm³ (20 °C)
Flash Pt: Does not flash
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=96870c784520df552dcde9a71610c883a1c71b50&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay GC (Product + Ethanol) Min. 99.8 %
Vapor Density 4.1 (vs air)
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.05 %
Refractive index (n 20/D) 1.444 - 1.446
Suitability for residue analysis Max. 25 ng/l
Contains ~1% ethanol as stabilizer
Vapor Pressure 160 mmHg ( 20 °C)
Refractive Index n20/D 1.445 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR