Sodium hydroxide solution 50%, Biotech Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS

2622-35EA 593.09 EUR
2622-35 2622-40 2622.9025PE 2622-SAMPLE 2622.9600
Sodium hydroxide solution 50%, Biotech Reagent, J.T.Baker®
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

Assay (NaOH) 49 - 51 %
Appearance (clear, colorless liquid) passes test
Potassium (K) max. 0.01 %
Identification (Na) passes test
Sodium Carbonate (Na2CO3) max. 1.0 %
Trace Impurities (in ppm) Chlorides (as Cl) max. 5
Trace Impurities (in ppm) Iron (Fe) max. 2
Trace Impurities (in ppm) Heavy Metals (as Pb) max. 1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR