High purity dNTPs
Supplier: GE HEALTHCARE
27-1850-04 27-1860-04 27-1870-04 27-1880-04
27-1850-04EA 478 EUR
27-1850-04 27-1860-04 27-1870-04 27-1880-04
High purity dNTPs
Nucleic Acid Reagents Nucleotides
dNTPs designed for applications requiring high purity such as amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling.
  • High purity dNTPs are functionally tested in long PCR to be PCR-qualified, and meet or exceed criteria for high-quality sequencing with thermo sequenase DNA polymerase
  • High purity (>98% triphosphate) confirmed by chromatographic analysis and ³¹P nuclear magnetic resonance (NMR) spectroscopy
Ordering information: Supplied as solids. For high purity solutions dNTPs, see illustra™ dNTP solutions.