MicroSpin™ DNA Clean-up, Columns
Supplier: GE HEALTHCARE
27-3565-01
27-3565-01EA 185 EUR
27-3565-01
MicroSpin™ DNA Clean-up, Columns
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
MicroSpin™ columns are designed for the rapid purification of DNA in conjunction with Sephadex™ G-25 DNA grade for use in a wide range of applications, including desalting, buffer exchange and removal of unincorporated nucleotides from end-labelled oligonucleotides.
  • For rapid buffer exchange/desalting of PCR products and other DNAs in a volume of 10 - 100 µl using spin-column and Sephadex™ G-25 DNA Grade (not included)
  • Excellent for rapid purification of newly synthesised oligonucleotides >10 mers in 100 to 150 µl of deprotection solution using spin column chromatography
  • Flexible for your experiments, allowing differing amounts of Sephadex™
  • For desalting/buffer exchange of DNA and removal of unincorporated radionucleotides from end-labelled oligonucleotides (at least 10 bases in length) in a volume of 10 to 100 µl
Excellent for rapid purification of newly synthesized oligonucleotides >10-mers in 100 to 150 µl of deprotection solution using spin-column chromatography. Can also be used for desalting/buffer exchange of DNA and removal of unincorporated radionucleotides from end-labeled oligonucleotides (at least 10 bases in length) in a volume of 10 to 100 µl.
They can be used for any DNA greater than 10 bases in length and are therefore highly suitable for the purification of oligonucleotides or very small DNA fragments following synthesis or a labeling reaction. For rapid buffer exchange/desalting of PCR products and other DNAs in a volume of 10 to 100 µl using spin-column chromatography and Sephadex G-25 DNA Grade (not included).