Microscale organic chemistry kit, Quickfit®
Supplier: SciLabware
ML1 ML/FE10 ML/FE25 ML/FP10 ML/FP25 ML/FR10 ML/C1 ML/C2 ML/FR25 ML/MF23 ML/RA3 ML/SH4 ML/ST
BIBBML1EA 535 EUR
BIBBML1 212-0618 212-0619 212-0620 212-0621 212-0622 201-3043 201-3036 212-0623 201-3061 271-0001 201-3007 201-3011
Microscale organic chemistry kit, Quickfit®
Microchemistry Kits
Microscale shemistry kit that is designed for organic chemistry experiments using milligram quantities of reagents. The components allow the construction of many assemblies including reflux, distillation, moisture sensitive reactions and fractional distillation.
  • Comprises 12 items of glassware that all feature 14/10 joint sizes for compatibility- see spare parts list
  • Other components compatible with the kit ar listed below under accessories
  • All components utilise a combination of standard Quickfit® ground glass joints together with screwthread couplings,
More Product Information