Sodium benzoate ≥99%, purified

Supplier: VWR Chemicals
Warning

27694.293EA 143 EUR
27694.293 27694.460
Sodium benzoate ≥99%, purified
Sodium benzoate
Formula: C₆H₅COONa
Melting Pt: 410…430 °C
Density: 1,44 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00012463
CAS Number: 532-32-1
EINECS: 208-534-8
REACH: 01-2119460683-35
Merck Index: 13,08654

Order Now

Specification Test Results

Assay (calculated on anhydrous) Min. 99 %
Heavy metals (as Pb) Max. 20 ppm
Loss on drying (100°C) Max. 1.0 %
Cl (Chloride) Max. 0.02 %
SO4 (Sulphate) Max. 0.03 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR