HiLoad Superdex columns, 200 PG
Supplier: Cytiva
28-9893-36 28-9893-35
28-9893-36EA 4231 EUR
28-9893-36 28-9893-35
HiLoad Superdex columns, 200 PG
Chromatography Columns
Superdex prep grade (pg) is produced by covalent bonding of dextran to highly cross-linked agarose.The separation properties of these media are determined by the dextran component
  • High resolution with short run times and good recovery
  • High capacity at high flow rates
  • Separations can be scaled up to production levels
  • Prepacked for convenience and reproducibility
  • 1/16" fittings for convenient connections to ÄKTA design systems
Steep selectivity curves give excellent resolving power for peptides and proteins in the molecular weight ranges, Mr < 10 000More Product Information