Cellulose Soxhlet extraction thimbles, Whatman™
Supplier: GE Healthcare - Whatman
2800-105 2800-185 2800-199 2800-226 2800-228 2800-250 2800-258 2800-266 2800-280 2800-282 2800-288 2800-300 2800-308 2800-330 2800-331 2800-338 2800-339 2800-373 2800-412 2800-432 2800-608 2810-228 2810-258 2810-338 2810-432 2810-339 2800-259 2800-260 2810-166 2810-266 2810-902
2800105.EA 124 EUR
2800105. 2800185.. 2800199. 734-2578 2800228. 2800250. 2800258. 2800266. 2800280. 2800282. 2800288. 2800300. 2800308. 2800330. 2800331. 2800338. 2800339. 2800373. 2800412. 2800432. 2800608. 2810228. 515-0168 2810338. 515-8943 537-0036 734-2579 734-2580 537-0034 537-0035 WHAT2810-902
Cellulose Soxhlet extraction thimbles, Whatman™
Extractors Extraction Thimbles
Cytiva's Whatman cellulose thimbles are manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems. Used in air and waste gas analysis.
  • Used in a wide variety of solid-liquid extraction studies, including lipid and pesticide analyses in food and soil samples
  • Suitable for temperatures up to 120 °C (248 °F)
  • Excellent mechanical strength and retention
  • Wall thickness of standard thimbles is 1 to 1,5 mm to support application need
  • Option for double thickness for added rigidity