Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger


Stabilization: 250-400 ppm Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

28552.290EA 50.6 EUR
28552.290 28552.324 28552.368 28552.461
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Tetrahydrofuran
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66
Melting Pt: –108,5 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3 II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=11aea9af60d6f59ae6301982bc1178f0ad862d59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %
Appearance Clear colourless liquid
IR Spectrum Passes test
Boiling point 65.0 - 66.5 °C
Density (20/4) 0.885 - 0.888
Free acidity Max. 0.0003 meq/g
Refractive index (20°C) 1.406 - 1.408
Peroxides (as H2O2) Max. 100 ppm
Water Max. 0.03 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR