Inlet liners, FocusLiner™, Supelco®
Supplier: Merck
2879805-U 2875705-U 2877001-U 2878605-U 2879201-U 2879205-U 2879501-U 2879505-U 2879525-U 2879601-U 2879605-U 2879625-U 2879825-U 2879901-U 2879905-U 2879925-U
2879805-UEA 225 EUR
2879805-U 2875705-U 2877001-U 2878605-U 2879201-U 2879205-U 2879501-U 2879505-U 2879525-U 2879601-U 2879605-U 2879625-U 2879825-U 2879901-U 2879905-U 2879925-U
Inlet liners, FocusLiner™, Supelco®
Chromatography Supplies Chromatography Inlet Liners
FocusLiner™ inlet liners are designed with wool plug to promote the rapid vaporisation of the entire sample, minimise mass discrimination, and prevent non-volatile material from entering the column. It incorporates a unique design to prevent the shifting of the wool plug during repeated injections or sudden inlet pressure changes. These liners are suitable for Agilent, Perkinelmer, Shimadzu, Finnigan, Varian and Thermo scientific equipped with split/splitless inlets.
  • Large surface area for maximum sample vaporisation
  • Wiping action removes residual liquid
More Product Information