Methanol, anhydrous ≥99.8% (by GC), Macron Fine Chemicals™

Supplier: MACRON AVANTOR BRAND
Danger

3004-45EA 179.87 EUR
3004-45 3004-25
Methanol, anhydrous ≥99.8% (by GC), Macron Fine Chemicals™
Methanol
Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=7bb8f9d74455604a6191a480ba063529ad0783c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (CH₃OH) (by GC) ≥99.8 %
Water (H₂O)(by Karl Fischer titrn) ≤0.1 %
Substances Reducing KMnO₄ Passes Test
Acidity Passes Test
Acetone Passes Test
Solubility in H₂O Passes Test
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR