Hydrofluoric acid ≥40%, Puriss. p.a. Reag. Ph. Eur., ISO analytical reagent, Fluka™

Supplier: HONEYWELL
Danger

30103-1L 30103-2.5L 30103-4X2.5L 30103-5L 30103-6X1L
30103-1LEA 128.64 EUR
30103-1L 30103-2.5L 30103-4X2.5L 30103-5L 30103-6X1L
Hydrofluoric acid ≥40%, Puriss. p.a. Reag. Ph. Eur., ISO analytical reagent, Fluka™
Hydrofluoric acid
Formula: HF
MW: 20.01 g/mol
Boiling Pt: 110 °C (1.013 hPa)
Melting Pt: –40 °C
Density: 1.140 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011346
CAS Number: 7664-39-3
EINECS: 231-634-8
UN: 1790
ADR: 8 II

Order Now

Specification Test Results

Assay Min. 40 %
Residue on ignition (SO₄) Max. 0.0005 %
Silver (Ag) Max. 0.000002 %
Aluminium (Al) Max. 0.000005 %
Arsenic (As) Max. 0.000005 %
Barium (Ba) Max. 0.00001 %
Beryllium (Be) Max. 0.000002 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000001 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00002 %
Germanium (Ge) Max. 0.000005 %
Potassium (K) Max. 0.00001 %
Lithium (Li) Max. 0.000002 %
Magnesium (Mg) Max. 0.00002 %
Manganese (Mn) Max. 0.000005 %
Molybdenum (Mo) Max. 0.000005 %
Sodium (Na) Max. 0.00002 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.000005 %
Strontium (Sr) Max. 0.000002 %
Titanium (Ti) Max. 0.00001 %
Thallium (Tl) Max. 0.000005 %
Vanadium (V) Max. 0.000005 %
Zinc (Zn) Max. 0.000005 %
Zirconium (Zr) Max. 0.00001 %
Ignition Residue ≤0.0005% (as SO₄)
Chloride (Cl) Max. 0.0001 %
Hexafluorosilicic acid (H₂SiF₆) Max. 0.005 %
Phosphate (PO₄) Max. 0.00005 %
Sulfate (SO₄) Max. 0.0002 %
Sulfite (SO₃) Max. 0.0002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR