Orthophosphoric acid ≥85%, Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Fluka™

Supplier: HONEYWELL
Danger

30417-1L 30417-2.5L 30417-4X2.5L 30417-6X1L 30417-1L-GL
30417-1LEA 100.92 EUR
30417-1L 30417-2.5L 30417-4X2.5L 30417-6X1L HONC30417-1L-GL
Orthophosphoric acid ≥85%, Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Fluka™
Orthophosphoric acid

Beilstein Number: 1921286

Formula: H₃PO₄
Boiling Pt: 158 °C (1,013 hPa)
Melting Pt: 21 °C
Density: 1,71 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III

Order Now

Specification Test Results

Assay Min. 85 %
Volatile acids (CH₃COOH) Max. 0.001 %
Water insoluble matter Max. 0.001 %
Arsenic (As) Max. 0.5 ppm
Calcium (Ca) Max. 0.001 %
Cadmium (Cd) Max. 0.0001 %
Cobalt (Co) Max. 0.0001 %
Chromium (Cr) Max. 0.0001 %
Copper (Cu) Max. 0.0001 %
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 0.0005 %
Magnesium (Mg) Max. 0.0005 %
Manganese (Mn) Max. 0.5 ppm
Sodium (Na) Max. 0.002 %
Nickel (Ni) Max. 0.0001 %
Lead (Pb) Max. 0.0001 %
Antimony (Sb) Max. 0.0002 %
Zinc (Zn) Max. 0.0002 %
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 2 ppm
Fluoride (F) Max. 0.0001 %
Nitrate (NO₃) Max. 3 ppm
Phosphite,Hypoph. (H₃PO₃) Max. 0.002 %
Phosphite, hypophosphite complying
Sulfate (SO₄) Max. 30 ppm
KMnO₄ reducing matter (as O) Max. 0.001 %
Substances precipitated by NH₃ complying
APHA Max. 10
Reducing impurities complying
Appearance of the solution complying
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR