Potassium chloride 99.0-100.5% (dried basis), crystals USP, FCC, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS
Warning

3046-01EA 71.69 EUR
3046-01 3046-05 3046-07 3046-R
Potassium chloride 99.0-100.5% (dried basis), crystals USP, FCC, J.T.Baker®
Potassium chloride

Avantor excipients are used during the formulation process in a wide range of applications. Avantor provides high-purity and performance excipients that serve as fillers, binders, disintegrants, lubicants, flavors, emulsifiers and preservatives. All the p

Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420 °C (1013 hPa)
Melting Pt: 773 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now

Specification Test Results

GMP Manufactured Product
Meets U.S.P Requirements
Food GMP Manufactured Product
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (KCl) (dried basis) 99.0 - 100.5 %
USP - Acidity or Alkalinity Passes Test
USP - Calcium and Magnesium Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Bromide (Br) ≤0.1 %
USP - Iodides (I) ≤0.005 %
USP - Loss on Drying ≤1.0 %
USP - Sodium (Na) Passes Test
FCC - Assay (KCl) (dried basis) 99.0 - 100.5 %
FCC - Acidity or Alkalinity Passes Test
FCC - Heavy Metals (as Pb) ≤5 ppm
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Iodide and/or Bromide Passes Test
FCC - Loss on Drying ≤1.0 %
FCC - Sodium (Na) Passes Test
Appearance (White crystals) Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR