Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM USP, Reag. Ph. Eur. volumetric solution

Supplier: VWR Chemicals
Warning

30472.297EA 95.7 EUR
30472.297 30472.322 30472.402 VWRS30472.323 30472.290
Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM USP, Reag. Ph. Eur. volumetric solution
Silver nitrate
Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444
Melting Pt: 210
Density: 1,01 g/cm3
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 3264
ADR: 8 III
Merck Index: 12,08661
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=166a8d8e5beb1527d4f3ab500692f0f7f86f3b10&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 0.0998 - 0.1002 mol/l
Conforms to Reag. Ph.Eur. Passes test
Conforms to USP Passes test
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR