Potassium sulphate ≥99.0%, fine crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS

3278.1000EA 70.32 EUR
3278.1000 3278-07 3278-R
Potassium sulphate ≥99.0%, fine crystals, BAKER ANALYZED® ACS, J.T.Baker®
Potassium sulphate
Formula: K₂SO₄
MW: 174.26 g/mol
Boiling Pt: 1698
Melting Pt: 1069
Density: 2.66 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011388
CAS Number: 7778-80-5
EINECS: 231-915-5
Merck Index: 13,07759

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements ≥99.0%
For Laboratory,Research,or Manufacturing Use ≤0.01%
Assay (K₂SO₄) ≥ 99.0 %
Insoluble Matter ≤ 0.01 %
pH of 5% Solution at 25°C 5.5 - 8.5
Chloride (Cl) ≤ 0.001 %
Calcium (Ca) ≤ 0.01 %
Magnesium (Mg) ≤ 0.005 %
Sodium (Na) ≤ 0.02 %
Nitrogen Compounds (as N) ≤ 5 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤ 5 ppm
Trace Impurities - Iron (Fe) ≤ 5 ppm
Meets Reagent Specifications for testing USP/NF monographs

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR