Flasks, round bottom with three necks and standard ground joints
Supplier: Lenz Laborglas GmbH & CO.KG
03303249 03303258 03303270 03304137 03304149 03304158 03304170 03304173 03304237 03304249 03304258 03304270 03304437 03304449 03304458 03304470 03304473 03304479 03304482 03305479 03305482 03304476 03305470 03305473 03305485 03305488 03304485 03304176 03305476 03303237 03303273 03304179 03304185 03304273 03304488 03305494
3303249.EA 91.7 EUR
3303249. 3303258. 3303270. 3304137. 3304149. 3304158. 3304170. 3304173. 3304237. 3304249. 3304258. 3304270. 3304437. 3304449. 3304458. 3304470. 3304473. 3304479. 3304482. 3305479. 3305482. 3304476. 3305470. 3305473. 3305485. 3305488. 3304485. 3304176. 3305476. 3303237. 3303273. 3304179. 3304185. 3304273. 3304488. 3305494.
Flasks, round bottom with three necks and standard ground joints
Flasks Round Bottom Flasks
DURAN®, borosilicate glass, with two parallel side necks, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Central neck and two parallel side necks. Matt white labelling area.
Order Now
SPECIFICATIONS
Environmentally Preferable