Aluminium oxide ≥99.7%, fused, insulating powder ∼ 60 µm

Supplier: ALFA AESAR

33200.A1 33200.A7
33200.A1EA 103 EUR
33200.A1 33200.A7
Aluminium oxide ≥99.7%, fused, insulating powder ∼ 60 µm
Aluminium oxide

CAS No.: 1344-28-1 Molecular Formula: Al2O3 Physical Form: ≈60 micron Odour: Odourless MDL No.: MFCD00003424


Used in dry filling for thermal insulation of electrical furnaces.

Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR