Supelco® PESTANAL® Organic Reference Standard, Mepanipyrim

Supplier: SUPELCO

33970-50MG
33970-50MGEA 141 EUR
33970-50MG
Supelco® PESTANAL® Organic Reference Standard, Mepanipyrim
Standards Organic Standards

  • GHS signal: GHS08, GHS09


Caution: P202 - P273 - P280 - P308 + P313 - P391 - P405

https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=982e558b9b50ee6c69c6ebfdeeab0d0933eec423&vid=982e558b9b50ee6c69c6ebfdeeab0d0933eec423&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Synonym(s) 4-Methyl-N-phenyl-6-(1-propynyl)-2-pyrimidinamine
Environmentally Preferable
CAS No. 110235-47-7
MDL No. MFCD01656050
Formula C₁₄H₁₃N₃
Molecular weight 223,27
Hazard class description H351 - H410
Flash point 131 °C
Shipping conditions STD
Storage conditions 2...30 °C
M-Clarity quality level MQ100

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR