Silver nitrate 0.0282 M AgNO3 (1 ml = 1 mg Cl) ≥99.5% volumetric solution, Fluka™

Supplier: HONEYWELL

35377-1L 35377-6X1L
35377-1LEA 80.8 EUR
35377-1L 35377-6X1L
Silver nitrate 0.0282 M AgNO3 (1 ml = 1 mg Cl) ≥99.5% volumetric solution, Fluka™
Silver nitrate
Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444
Melting Pt: 210
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
Merck Index: 12,08661
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=166a8d8e5beb1527d4f3ab500692f0f7f86f3b10&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Factor 0.999 - 1.001
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR