Tin(II) chloride, ultra dry ≥99.9985% (metals basis)

Supplier: ALFA AESAR
Danger


Ampuled under argon

35723.03 35723.06 35723.22 35723.14
35723.03EA 95.2 EUR
35723.03 35723.06 35723.22 35723.14
Tin(II) chloride, ultra dry ≥99.9985% (metals basis)
Tin (II) chloride

Purity: 99.9985% CAS No.: 7772-99-8 Molecular Formula: SnCl2 Formula Weight: 189.62 Physical Form: Powder, Ampouled under argon Density: 3.95 Melting Point: 247° Boiling Point: 652° Merck Reference: 14,8783 UN No.: 3260 MDL No.: MFCD00011241

Formula: SnCl₂
MW: 189.62 g/mol
Boiling Pt: 652 °C
Melting Pt: 247 °C
Density: 3.95
Storage Temperature: Ambient
MDL Number: MFCD00011241
CAS Number: 7772-99-8
EINECS: 231-868-0
UN: 3260
ADR: 8 III
Merck Index: 14,8783

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR