Magnesium ≥99.6% (metals basis excluding Ca), powder -100+200 mesh

Supplier: ALFA AESAR
Danger

36194.30
36194.30EA 157 EUR
36194.30
Magnesium ≥99.6% (metals basis excluding Ca), powder -100+200 mesh
Magnesium

Purity: 99.6% CAS No.: 7439-95-4 Physical Form: Ca typically 0.1-1% Appearance: White Odour: Odourless UN No.: 1418 MDL No.: MFCD00085308


For vacuum deposition

Formula: Mg
MW: 24.3 g/mol
Boiling Pt: 1107
Melting Pt: 651
Density: 1.74 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1418
ADR: 4.3 II
Merck Index: 13,05675

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR