Ytterbium(III) oxide ≥99.9%

Supplier: ALFA AESAR

36252.14 36252.22
36252.14EA 247 EUR
36252.14 36252.22
Ytterbium(III) oxide ≥99.9%
Ytterbium (III) oxide

Purity: 99.9% CAS No.: 1314-37-0 Molecular Formula: Yb2O3 Formula Weight: 394.08 Physical Form: 3-12mm Sintered Lump Density: 9.2 Melting Point: 2227° Merck Reference: 14,10105 MDL No.: MFCD00011290

Formula: O₃Yb₂
MW: 394.08 g/mol
Melting Pt: 2227 °C
Density: 9.2
Storage Temperature: Ambient
MDL Number: MFCD00011290
CAS Number: 1314-37-0
EINECS: 215-234-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR