4-Chlorophenylmagnesium bromide 1 M in THF/toluene, AcroSeal®

Supplier: ACROS ORGANICS
Danger

377251000 377258000
377251000.EA 82 EUR
377251000. 377258000.
4-Chlorophenylmagnesium bromide 1 M in THF/toluene, AcroSeal®
4-Chlorophenylmagnesium bromide
Formula: C₆H₄BrClMg
Density: 1 g/cm³
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD00009926
CAS Number: 873-77-8
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Yellow to green to brown Slightly hazy solution
Titration Active base 0.95 to 1.15 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR