Diisobutylaluminium chloride 0.8 M in heptane, AcroSeal®

Supplier: ACROS ORGANICS
Danger

377561000 377568000
377561000.EA 106 EUR
377561000. 377568000.
Diisobutylaluminium chloride 0.8 M in heptane, AcroSeal®
Diisobutylaluminium chloride
Formula: C₈H₁₈AlCl
Density: 0,726 g/cm³
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008927
CAS Number: 1779-25-5
EINECS: 217-216-8
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Clear colorless Solution
Strength 0.78 to 0.86 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR