Sodium dihydrogen phosphate monohydrate 98.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS

3818-01EA 205.81 EUR
3818-01 3818-05 3818-07
Sodium dihydrogen phosphate monohydrate 98.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Sodium dihydrogen phosphate monohydrate
Formula: NaH₂PO₄·H₂O
Melting Pt: 100 °C
Density: 2,04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149208
CAS Number: 10049-21-5
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 13,08734
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=5597299de719468db5a68462cc750045659b46af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements 98.0 - 102.0%
For Laboratory,Research,or Manufacturing Use 4.1 - 4.5
Assay (NaH₂PO₄·H₂O) 98.0 - 102.0 %
pH of 5% Solution at 25°C 4.1 - 4.5
Insoluble Matter ≤0.01 %
Chloride (Cl) ≤5 ppm
ACS - Sulfate (SO₄) ≤0.003 %
Calcium (Ca) ≤0.005 %
Potassium (K) ≤0.01 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR