N,N'-Ditridecyl-3,4,9,10-perylenetetracarboxylic diimide 95%, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY


MQ100

383783-1G
383783-1GEA 270 EUR
383783-1G
N,N'-Ditridecyl-3,4,9,10-perylenetetracarboxylic diimide 95%, Sigma-Aldrich®
N,N'-Ditridecyl-3,4,9,10-perylenetetracarboxylic diimide
Formula: C₅₀H₆₂N₂O₄
MW: 755.06 g/mol
Melting Pt: >300 °C (lit.)
Storage Temperature: Room Temperature
MDL Number: MFCD00134587
CAS Number: 95689-92-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a70219025bbf989f3dda8cb48491f60540fc6eb&vid=8a70219025bbf989f3dda8cb48491f60540fc6eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Colour) Dark Red to Brown Conforms to Requirements
Appearance (Form) Powder or Crystals or Fibers
Infrared Spectrum Conforms to Structure
UV/VIS Absorbance in Toluene 523 - 529 nm
Carbon 74.7 - 84.3%
Nitrogen 3.4 - 3.9%
Purity (TLC) ≥95%
Solubility (Turbidity), c = 10 mg/ml; Toluene Clear to Turbid
Solubility (Colour) Brown to Very Dark Brown and Dark Red


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR