Aluminium oxide substrate 10x10x0.5 mm, polished one side R plane

Supplier: ALFA AESAR

38500.KT
38500.KTEA 0 EUR
38500.KT
Aluminium oxide substrate 10x10x0.5 mm, polished one side R plane
Aluminium oxide

Molecular Formula: Al2O3 MDL No.: MFCD00003424

Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980
Melting Pt: 2045
Density: 3.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR