Aluminium lanthanum (III) oxide substrate 10x10x0.5 mm, polished one side 100 orientation

Supplier: ALFA AESAR

38501.KT 38501.KU
38501.KTEA 197 EUR
38501.KT 38501.KU
Aluminium lanthanum (III) oxide substrate 10x10x0.5 mm, polished one side 100 orientation
Aluminium lanthanum (III) oxide

CAS No.: 12003-65-5 Molecular Formula: LaAlO3 Odour: Odourless MDL No.: MFCD00144550

Formula: AlLaO₃
MW: 213.89 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00144550
CAS Number: 12003-65-5
EINECS: 234-433-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR