Silicon carbide ≥99% (metals basis), whiskers

Supplier: ALFA AESAR
Warning

38787.09 38787.18 38787.30
38787.09EA 92.8 EUR
38787.09 38787.18 38787.30
Silicon carbide ≥99% (metals basis), whiskers
Silicon carbide

Purity: 99% CAS No.: 409-21-2 Molecular Formula: SiC Formula Weight: 40.10 Physical Form: 1.5 micron dia x approx 18 micron length Density: 3.2 Melting Point: 2700°C Merck Reference: 14,8492 MDL No.: MFCD00049531


Abrasive, polishing, grinding

Formula: SiC
MW: 40.1 g/mol
Melting Pt: 2700 °C
Density: 3.2
Storage Temperature: Ambient
MDL Number: MFCD00049531
CAS Number: 409-21-2
EINECS: 206-991-8
Merck Index: 14,8492
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bd532b391b9c28edcb8adac5fe5ed958ccaa487&vid=0bd532b391b9c28edcb8adac5fe5ed958ccaa487&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR