Compact incubator with forced convection, Heratherm®
Supplier: THERMO BRANDED
50125882 50140500
390-0466EA 986 EUR
390-0466 390-1299
Compact incubator with forced convection, Heratherm®
Incubators Compact Benchtop Incubators
Heratherm® Compact incubator features forced convection providing excellent temperature uniformity and stability, and fast recovery times after opening the door. This unit is recommended for use as a personal work space on the benchtop, and for BOD/water pollution testing. The unit is made of corrosion-free, easy to clean moulded polymer with rounded corners. An internal light and window in door facilitate sample observation.
  • Easy to use control interface
  • Display resolution 1 °C
  • Minimal bench space needed - ideal for space-restricted laboratory areas
  • Easy
More Product Information