Heracell™ VIOS CO₂ incubators
Supplier: Heraeus
51900293 51901126 51901144 51030704 51030661 51031540 51031823 51033548 51033549 51033558 51033559 51033588 51033589 51033598 51033599 51033571 51033575 50163006 50163007 50163008 50163009
390-4426EA 2 EUR
390-4426 HERA51901126 390-0906 390-0898 390-0900 390-1038 390-1040 390-1526 390-1527 390-1528 390-1529 390-1532 390-1533 390-1534 390-1535 390-1536 390-1539 390-1540 390-1541 390-1542 390-1543
Heracell™ VIOS CO₂ incubators
Incubators CO₂ Incubators
The Heracell™ VIOS series has been designed for sensitive cultures like stem and primary cells in leading research, pharmaceutical and clinical applications. The units are available with either electropolished stainless steel or 100% pure copper interiors and have adjustable, perforated shelving, easy to clean corners with convenient access port and a reversible exterior door for added flexibility.
  • THRIVE active airflow maintains homogeneous conditions avoiding unwanted sample variation with fast recovery times of under 10 minutes utilising long life,
More Product Information