CO₂ incubators with hot air sterilisation, CB series
Supplier: Binder
9640-0009 9640-0010 9640-0013 9640-0005 9640-0014 9640-0027
390-1616EA 12940 EUR
390-1616 390-1617 390-1620 390-1624 390-1621 390-1672
CO₂ incubators with hot air sterilisation, CB series
Incubators CO₂ Incubators
CO₂ incubators with a built in CO₂ sensor which can be sterilised. Units offer optimum growth conditions for cell cultures and meet demanding requirements for best possible cell growth and maximum sample safety. The ANTI.PLENUM design minimises surfaces due to the seamless, deep-drawn inner chamber without fixtures and featuring an integrated shelf support system. This reduces the risk of contamination, giving less surface area and fewer parts to clean manually, and removes the running costs of consumables. Reliable decontamination is provided by the standard-compliant 180 °C hot airMore Product Information