Rolled flange bottles, narrow mouth, without clamp closure, DURAN®
Supplier: DWK Life Sciences
214612403
391-0025EA 19.9 EUR
391-0025
Rolled flange bottles, narrow mouth, without clamp closure, DURAN®
Bottles Cell Culture Bottles
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For sampling and cultivation.