Silicone stoppers
Supplier: HIRSCHMANN
8905510 8905512 8905515 8905517 8905519 8905522 8905524 8905532 8905542 8905552 8905612 8905617 8905622 8905720 8905730 8905740 8905755 8905828 8905835 8905840 8905912 8905915 8905917 8905919 8905922 8905924 8905932 8905942 8905952 8905765 8905928
391-1969EA 35.6 EUR
391-1969 391-0139 391-1957 391-0141 391-0142 391-1956 391-0143 391-1955 391-0144 391-1961 391-1954 391-0146 391-0147 391-1953 391-1952 391-1960 391-1951 391-1950 391-1949 391-1962 391-1963 391-1964 391-1965 391-1966 391-1959 391-1958 391-1967 391-1968 217-0835 HIRS8905765 HIRS8905928
Silicone stoppers
Stoppers Silicone Stoppers
SILICOSEN® and BIO-SILICO® sterile stoppers are manufactured in a special process and have regular pores, thus making them ideal for the preparation and sterilisation of culture media. The uniform pores and good permeability to air make the stoppers ideally suitable for cultivation of aerobic microorganisms.
  • Chemical and temperature resistant
  • Liquid repellent
  • Reusable
SILICOSEN® T-type and L-type: Low humidity evaporation reduces dehydration of cultures.
SILICOSEN® C-type: Excellent permeability and secure fitting, thanks to seal lip. Ideally suitableMore Product Information