N,N-Dimethylformamide ≥99.8% ACS

Supplier: ALFA AESAR
Danger

39117.AP 39117.K2 39117.K7 39117.M1 39117.M6
39117.APEA 52.5 EUR
39117.AP 39117.K2 39117.K7 39117.M1 39117.M6
N,N-Dimethylformamide ≥99.8% ACS
N,N-Dimethylformamide
Formula: HCON(CH₃)₂
Boiling Pt: 153 °C
Density: 0,944
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=fb9c58451426a2acf92f6525238f8198a167f088&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR