Nickel hydroxyacetate (27 - 29% Ni)

Supplier: ALFA AESAR
Danger

39456.14
39456.14EA 0 EUR
39456.14
Nickel hydroxyacetate (27 - 29% Ni)
Nickel hydroxyacetate

CAS No.: 41587-84-2 Molecular Formula: C4H6NiO6 Formula Weight: 208.78 Physical Form: Powder Appearance: Green UN No.: UN3077 MDL No.: MFCD00070414

Formula: C₄H₆NiO₆
MW: 208.78 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070414
CAS Number: 41587-84-2
UN: 3077
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR