Sample splitters, RT 6,5 - RT 75
Supplier: Retsch GmbH
40.610.0002 40.610.0003 40.610.0001 40.610.0004 40.610.0005 40.610.0006
40.610.0002EA 1050 EUR
40.610.0002 40.610.0003 40.610.0001 40.610.0004 40.610.0005 40.610.0006
Sample splitters, RT 6,5 - RT 75
Sample Dividers
Sample splitters are used for the simple dividing and reduction of bulk materials of all kinds. Ideal for the on site reduction of sample material. Depending on the particle size, material and particle size distribution, the opening width of the passage should be 2,5 to 3 times greater than the diameter of the largest particle (particle size factor).
  • Easy to use and easy to clean
  • Consist of one dividing head, one stand and three receptacles
  • Can be used in the laboratory and on site
  • High precision manual dividing process
Ordering information: RT 6,5More Product Information