Jaw crushers, BB 50
Supplier: Retsch GmbH
20.062.0001 20.062.0005 20.062.0004 20.062.0003 20.062.0002
412-0215EA 22490 EUR
412-0215 412-0219 412-0218 412-0217 412-0216
Jaw crushers, BB 50
Mills
The BB 50 is a robust and powerful forced feed crusher that has been specifically designed for rapid but gentle, bench-top sample preparation of hard, brittle and hard-ductile materials in batches. The dust-tight, laboratory instrument is suitable for applications including constructional materials, mineralogy, metallurgy, ceramics and environmental analysis. A convenient operating keypad with digital display and a memory function for gap width setting, facilitates reproducible operation.
  • Final fineness down to 0,5 mm *
  • Breaking jaws and wearing plates
More Product Information