Polyvinyl alcohol, M.W. ∼ 10,000-26,000 98-99%, low molecular weight, hydrolysed

Supplier: ALFA AESAR

41241.14 41241.22 41241.36
41241.14EA 24.1 EUR
41241.14 41241.22 41241.36
Polyvinyl alcohol, M.W. ∼ 10,000-26,000 98-99%, low molecular weight, hydrolysed
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR