Antimony standard solution, 1000 mg/L Sb in water with nitric acid and tartaric acid (each max. 1%) (from Sb), standard for ICP, Specpure®

Supplier: ALFA AESAR

41682.AD 41682.AE 41682.AP
41682.ADEA 63.4 EUR
41682.AD 41682.AE 41682.AP
Antimony standard solution, 1000 mg/L Sb in water with nitric acid and tartaric acid (each max. 1%) (from Sb), standard for ICP, Specpure®
Standards Cationic Standards Antimony Standards

Molecular Formula: Sb in H₂O/tartaric acid/tr. HNO₃
Physical Form: Liquid
MDL No.: MFCD00134030


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR