Copper(II) hexafluoroacetylacetonate hydrate ≥99.99% (metals basis)

Supplier: ALFA AESAR
Warning

41870.02 41870.04 41870.09
41870.02EA 0 EUR
41870.02 41870.04 41870.09
Copper(II) hexafluoroacetylacetonate hydrate ≥99.99% (metals basis)
Copper (II) hexafluoroacetylacetonate hydrate

Purity: 99.99% CAS No.: 155640-85-0 Molecular Formula: Cu(CF3COCHCOCF3)2.xH2O Formula Weight: 477.64(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Boiling Point: 220° dec. MDL No.: MFCD00151019

Formula: C₁₀H₂CuF₁₂O₄·H₂O
MW: 477.64 g/mol (anhy)
Boiling Pt: 220 °C (dec.)
Melting Pt: ∼ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00151019
CAS Number: 155640-85-0
EINECS: 238-850-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR