Hydrofluoric acid 48 - 51% in water ACS

Supplier: ACROS ORGANICS
Danger

423800250 423805000 423800025
423800250.EA 44.3 EUR
423800250. 423805000. 423800025.
Hydrofluoric acid 48 - 51% in water ACS
Hydrofluoric acid
Formula: HF
Melting Pt: –35 °C
Density: 1,15 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011346
CAS Number: 7664-39-3
EINECS: 231-634-8
UN: 1790
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Residue after ignition ≤0.0005 %
Titration with NaOH 48.0 to 51.0 %
Chloride (Cl) ≤5 ppm
Phosphate (PO4) ≤1 ppm
SO4 and SO3 (as SO4) ≤5 ppm
Arsenic (As) ≤0.05 ppm
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤1 ppm
Heavy metals (as Pb) ≤0.5 ppm
Fluorosilicic acid ≤0.01 % (H2SiF6)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR