Palladium ≥99.95% (metals basis), sponge , -100/+325 mesh

Supplier: ALFA AESAR
Warning

42431.03 42431.06 42431.14
42431.03EA 382 EUR
42431.03 42431.06 42431.14
Palladium ≥99.95% (metals basis), sponge , -100/+325 mesh
Palladium

Purity: 99.95% CAS No.: 7440-05-3 Appearance: Dark grey to black Odour: Odourless UN No.: 3089 MDL No.: MFCD00011167

Formula: Pd
MW: 106.420 g/mol
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
UN: 3089
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR