Thiocarbonyl chloride 85%, AcroSeal®

Supplier: ACROS ORGANICS
Danger

426441000
426441000.EA 150 EUR
426441000.
Thiocarbonyl chloride 85%, AcroSeal®
Thiocarbonyl chloride
Formula: CCl₂S
Flash Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004918
CAS Number: 463-71-8
EINECS: 207-341-6
UN: 2474
ADR: 6.1,I

Order Now

Specification Test Results

Appearance Orange-red to red Liquid
GC ≥97.5 %
Refractive index 1.4025 to 1.4065 (20°C, 589 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR