Rhodium ≥99.9% (metals basis), sponge -20 mesh

Supplier: ALFA AESAR
Warning

42866.03 42866.06 42866.09
42866.03EA 2740 EUR
42866.03 42866.06 42866.09
Rhodium ≥99.9% (metals basis), sponge -20 mesh
Rhodium

Purity: 99.9% CAS No.: 7440-16-6 Odour: Odourless UN No.: 3089 MDL No.: MFCD00011201

Formula: Rh
Storage Temperature: Ambient
MDL Number: MFCD00011201
CAS Number: 7440-16-6
EINECS: 231-125-0
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR