Copper ≥99.9999% (metals basis), foil, Puratronic®, Thickness: 1.0 mm (0.04 in)

Supplier: ALFA AESAR

42976.FF 42976.FI 42976.GH
42976.FFEA 113 EUR
42976.FF 42976.FI 42976.GH
Copper ≥99.9999% (metals basis), foil, Puratronic®, Thickness: 1.0 mm (0.04 in)
Copper

Purity: 99.9999% CAS No.: 7440-50-8 Physical Form: ≈5.6g/25x25mm Odour: Odourless MDL No.: MFCD00010965

Formula: Cu
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8,94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR