Digadolinium trioxide ≥99.99% (REO, rare earth oxide basis), nanopowder

Supplier: ALFA AESAR
Warning

44020.14 44020.22 44020.36
44020.14EA 190 EUR
44020.14 44020.22 44020.36
Digadolinium trioxide ≥99.99% (REO, rare earth oxide basis), nanopowder
Digadolinium trioxide
Formula: Gd₂O₃
Density: 7,407
Storage Temperature: Ambient
MDL Number: MFCD00011026
CAS Number: 12064-62-9
EINECS: 235-060-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR