Reaction block systems for RH basic, RH digital, RCT basic, RET basic and RET control-visc magnetic hotplate stirrers
Supplier: IKA
25000627 25000628 25000629 25000630 25000631 25000632 25000633 25000634 25000636 25000637 25000626 25000639 25000641 25000642 25000644 25000645 25000832 25000833 25000834 25000638 20007954 25003280
442-0977EA 113 EUR
442-0977 442-0978 442-0973 442-0974 442-0975 442-0969 442-0970 442-0984 442-0962 442-0963 442-0976 442-0983 442-0966 442-0964 442-0967 442-0965 442-0985 442-0982 442-0981 442-0968 IKAA20007954 IKAA25003280
Reaction block systems for RH basic, RH digital, RCT basic, RET basic and RET control-visc magnetic hotplate stirrers
Block Heaters Blocks
Reaction blocks allow syntheses to be carried out in round-bottomed flasks at temperatures up to 180 °C, avoiding the use of oil baths. The system ensures optimum heat transfer from the heating plate directly into the medium. Uniform mixing is also guaranteed since there is no interference to the magnetic field from the aluminium blocks. Reaction blocks range from 100 to 5000 ml in size, and inserts accommodating round bottom flasks from 10 to 3000 ml are available. Due to the wide range of interchangeable inserts, this system can be used for a range of different applications with the RCT,More Product Information