Polyethylene glycol 6,000 for biochemistry

Supplier: VWR Chemicals

442714KEA 87.3 EUR
442714K
Polyethylene glycol 6,000 for biochemistry
Polyethylene glycol 6,000
Formula: H(OCH₂CH₂)nOH
MW: 6000 g/mol
Boiling Pt: >250 °C
Melting Pt: 60…63 °C
Density: 1.21 g/cm³ (20 °C)
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD01779614
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Specification Test Results

IR Spectrum Passes test
Average molecular weight 5000 - 7000
Heavy metals (as Pb) Max. 10 ppm
Absorbance (260 nm) (0,01 mol/l) Max. 0.2
Absorbance (280 nm) (0,01 mol/l) Max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR